ENAH303683A6 Enamine

5-(furan-2-yl)imidazolidine-2,4-dione

  • MDL number MFCD00159525
  • Cas Number : 79003-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-147
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1255200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1255200
Availability details for
Quantity
  • Availability
  • Add to Cart