ENAH303683AE Enamine

methyl 2-hydroxy-2,3-dimethylbutanoate

  • MDL number MFCD16769963
  • Cas Number : 856293-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1255958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1255958
Availability details for
Quantity
  • Availability
  • Add to Cart