ENAH303683B0 Enamine

3-cyclopropyl-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD16707040
  • Cas Number : 1249837-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1256709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1256709
Availability details for
Quantity
  • Availability
  • Add to Cart