ENAH303683B3 Enamine

2-amino-3-(dimethylamino)-2-methylpropanoic acid

  • MDL number MFCD16136254
  • Cas Number : 1249298-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1258371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1258371
Availability details for
Quantity
  • Availability
  • Add to Cart