ENAH303683B5 Enamine

2-chloro-6-ethyl-5-methylpyridine-3-carbonitrile

  • MDL number MFCD10478932
  • Cas Number : 108244-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-78
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1259097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1259097
Availability details for
Quantity
  • Availability
  • Add to Cart