ENAH303683C1 Enamine

oxane-4-carbonyl isothiocyanate

  • MDL number MFCD12189012
  • Cas Number : 1153207-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1262234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1262234
Availability details for
Quantity
  • Availability
  • Add to Cart