ENAH303683CD Enamine

2-amino-2-(2,3-dichlorophenyl)acetic acid hydrochloride

  • MDL number MFCD04966781
  • Cas Number : 1025447-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1265433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1265433
Availability details for
Quantity
  • Availability
  • Add to Cart