ENAH303683D2 Enamine

1-cyclopentyl-4,5,6,7-tetrahydro-1H-indazol-4-amine

  • MDL number MFCD21221268
  • Cas Number : 1484115-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1265954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1265954
Availability details for
Quantity
  • Availability
  • Add to Cart