ENAH30368440 Enamine

1-bromo-5-(tert-butoxy)pentane

  • MDL number MFCD19676922
  • Cas Number : 169054-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1601481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1601481
Availability details for
Quantity
  • Availability
  • Add to Cart