ENAH30368456 Enamine

1-(3-chlorophenyl)-2-fluoroethan-1-amine hydrochloride

  • MDL number MFCD28044277
  • Cas Number : 1639867-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1605496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1605496
Availability details for
Quantity
  • Availability
  • Add to Cart