ENAH30368457 Enamine

1,4-diazabicyclo[2.2.2]octane-2-carboxylic acid dihydrochloride

  • MDL number MFCD20921501
  • Cas Number : 69491-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1605566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1605566
Availability details for
Quantity
  • Availability
  • Add to Cart