ENAH3036845B Enamine

2-(2,6-dimethoxyphenyl)pyrrolidine hydrochloride

  • MDL number MFCD31719676
  • Cas Number : 2253632-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1606097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1606097
Availability details for
Quantity
  • Availability
  • Add to Cart