ENAH3036845D Enamine

5-azaspiro[3.4]octan-7-ol hydrochloride

  • MDL number MFCD31718533
  • Cas Number : 2247102-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1608049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1608049
Availability details for
Quantity
  • Availability
  • Add to Cart