ENAH30368462 Enamine

3-amino-5,7-dimethyladamantane-1-carboxylic acid

  • Cas Number : 14664-67-6
  • Molecular Weight : 223.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-5,7-dimethyl-1-adamantanecarboxylic acid
Inchi Code 1S/C13H21NO2/c1-10-3-11(2)5-12(4-10,9(15)16)8-13(14,6-10)7-11/h3-8,14H2,1-2H3,(H,15,16)
Inchi Key VGTIAGCVYBEACG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1608894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1608894
Availability details for
Quantity
  • Availability
  • Add to Cart