ENAH30368464 Enamine

2-(3-methylphenyl)propanenitrile

  • MDL number MFCD11036538
  • Cas Number : 75920-44-4
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3-methylphenyl)propanenitrile
Inchi Code 1S/C10H11N/c1-8-4-3-5-10(6-8)9(2)7-11/h3-6,9H,1-2H3
Inchi Key RMGYNPBCMHKZCJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1610734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1610734
Availability details for
Quantity
  • Availability
  • Add to Cart