ENAH30368465 Enamine

4-chloro-6-(4-ethyl-3-methylpiperazin-1-yl)pyrimidine

  • MDL number MFCD18847834
  • Cas Number : 1342904-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1265992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1265992
Availability details for
Quantity
  • Availability
  • Add to Cart