ENAH3036846E Enamine

4-[(tert-butoxy)carbonyl]-3,4-dihydro-2H-oxazine-3-carboxylic acid

  • MDL number MFCD31718536
  • Cas Number : 2248257-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1654370

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1654370
Availability details for
Quantity
  • Availability
  • Add to Cart