ENAH30368470 Enamine

6-fluoro-2-methyl-3-nitrobenzoic acid

  • MDL number MFCD28347722
  • Cas Number : 1079992-96-3
  • Molecular Weight : 199.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-2-methyl-3-nitrobenzoic acid
Inchi Code 1S/C8H6FNO4/c1-4-6(10(13)14)3-2-5(9)7(4)8(11)12/h2-3H,1H3,(H,11,12)
Inchi Key DEPIFWRMRCNBGQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1654914

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1654914
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart