ENAH30368471 Enamine

6-bromo-2-methoxy-1,3-benzothiazole

  • MDL number MFCD22491570
  • Cas Number : 1805217-96-2
  • Molecular Weight : 244.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-2-methoxybenzo[d]thiazole
Inchi Code 1S/C8H6BrNOS/c1-11-8-10-6-3-2-5(9)4-7(6)12-8/h2-4H,1H3
Inchi Key NWJQRJRIBQOMCI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1654947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1654947
Availability details for
Quantity
  • Availability
  • Add to Cart