ENAH30368472 Enamine

methyl 3-carbamothioylpropanoate

  • MDL number MFCD19207749
  • Cas Number : 80592-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1655702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1655702
Availability details for
Quantity
  • Availability
  • Add to Cart