ENAH30368473 Enamine

3a-methyl-1,1-dioxo-hexahydro-1lambda6-pyrrolo[1,2-b][1,2]thiazole-2-carboxylic acid

  • MDL number MFCD31736284
  • Cas Number : 2247102-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1655962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1655962
Availability details for
Quantity
  • Availability
  • Add to Cart