ENAH3036847B Enamine

1,4-dioxadispiro[4.1.4^{7}.3^{5}]tetradecan-12-ol

  • MDL number MFCD31736287
  • Cas Number : 1628927-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1662351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1662351
Availability details for
Quantity
  • Availability
  • Add to Cart