ENAH3036847F Enamine

(2S)-5-bromo-2,3-dihydro-1H-indole-2-carboxylic acid

  • MDL number MFCD21341265
  • Cas Number : 1344389-49-6
  • Molecular Weight : 242.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-5-bromo-2,3-dihydro-1H-indole-2-carboxylic acid
Inchi Code 1S/C9H8BrNO2/c10-6-1-2-7-5(3-6)4-8(11-7)9(12)13/h1-3,8,11H,4H2,(H,12,13)/t8-/m0/s1
Inchi Key JOMWUCRGBBSIIL-QMMMGPOBSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1268486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1268486
Availability details for
Quantity
  • Availability
  • Add to Cart