ENAH3036848A Enamine

6-chloro-4-hydroxypyridine-2-carboxylic acid

  • MDL number MFCD13189291
  • Cas Number : 1060809-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1270186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1270186
Availability details for
Quantity
  • Availability
  • Add to Cart