ENAH3036848B Enamine

{6,6-dimethylspiro[3.3]heptan-2-yl}methanesulfonyl chloride

  • MDL number MFCD31718860
  • Cas Number : 2167263-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1270333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1270333
Availability details for
Quantity
  • Availability
  • Add to Cart