ENAH3036848D Enamine

1-[(tert-butoxy)carbonyl]-4-(fluoromethyl)piperidine-4-carboxylic acid

  • MDL number MFCD28615471
  • Cas Number : 1785107-71-2
  • Molecular Weight : 261.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(tert-butoxycarbonyl)-4-(fluoromethyl)piperidine-4-carboxylic acid
Inchi Code 1S/C12H20FNO4/c1-11(2,3)18-10(17)14-6-4-12(8-13,5-7-14)9(15)16/h4-8H2,1-3H3,(H,15,16)
Inchi Key MDSRZVVMJRJURG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1664086

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1664086
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart