ENAH30368499 Enamine

4-chloro-N-(2-methylpropyl)pyrimidin-2-amine

  • MDL number MFCD20688298
  • Cas Number : 1201176-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1665897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1665897
Availability details for
Quantity
  • Availability
  • Add to Cart