ENAH303684A1 Enamine

4-bromo-2-chloro-5-methylaniline

  • MDL number MFCD20233409
  • Cas Number : 888485-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1272189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1272189
Availability details for
Quantity
  • Availability
  • Add to Cart