ENAH303684AD Enamine

1-(difluoromethyl)-2,3-difluoro-5-nitrobenzene

  • MDL number MFCD28790630
  • Cas Number : 1804516-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1272919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1272919
Availability details for
Quantity
  • Availability
  • Add to Cart