ENAH303684AE Enamine

2-methyl-5-[(2S)-pyrrolidin-2-yl]-1,3,4-oxadiazole

  • MDL number MFCD30828780
  • Cas Number : 1646783-30-3
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-2-methyl-5-(pyrrolidin-2-yl)-1,3,4-oxadiazole
Inchi Code 1S/C7H11N3O/c1-5-9-10-7(11-5)6-3-2-4-8-6/h6,8H,2-4H2,1H3/t6-/m0/s1
Inchi Key BLXQYIORYQVKFE-LURJTMIESA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1669831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1669831
Availability details for
Quantity
  • Availability
  • Add to Cart