ENAH303684AF Enamine

2-methyl-5-[(2R)-pyrrolidin-2-yl]-1,3,4-oxadiazole

  • MDL number MFCD31719677
  • Cas Number : 2253619-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1669841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1669841
Availability details for
Quantity
  • Availability
  • Add to Cart