ENAH303684C5 Enamine

2,3,6-trifluoro-4-(trifluoromethyl)benzoic acid

  • MDL number MFCD03701073
  • Molecular Weight : 244.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3,6-trifluoro-4-(trifluoromethyl)benzoic acid
Inchi Code 1S/C8H2F6O2/c9-3-1-2(8(12,13)14)5(10)6(11)4(3)7(15)16/h1H,(H,15,16)
Inchi Key QWOCVHZTKMSWRC-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1387031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1387031
Availability details for
Quantity
  • Availability
  • Add to Cart