ENAH303684CC Enamine

4,4,5,5-tetramethyl-2-(1-methylnaphthalen-2-yl)-1,3,2-dioxaborolane

  • MDL number MFCD28347757

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1425817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1425817
Availability details for
Quantity
  • Availability
  • Add to Cart