ENAH303684E2 Enamine

2,2-dimethyl-6,8-dioxa-7lambda6-thiaspiro[3.5]nonane-7,7-dione

  • MDL number MFCD31718883
  • Cas Number : 2248349-54-2
  • Molecular Weight : 206.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-dimethyl-6,8-dioxa-7-thiaspiro[3.5]nonane 7,7-dioxide
Inchi Code 1S/C8H14O4S/c1-7(2)3-8(4-7)5-11-13(9,10)12-6-8/h3-6H2,1-2H3
Inchi Key VMCJBPYNXYVTCY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1678122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1678122
Availability details for
Quantity
  • Availability
  • Add to Cart