ENAH303684E3 Enamine

spiro[2.5]octane-1-carbaldehyde

  • MDL number MFCD24683914
  • Cas Number : 200055-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1679641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1679641
Availability details for
Quantity
  • Availability
  • Add to Cart