ENAH303684E5 Enamine

5-oxaspiro[2.5]octane-1-carbaldehyde

  • MDL number MFCD28118086
  • Cas Number : 1784642-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1679673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1679673
Availability details for
Quantity
  • Availability
  • Add to Cart