ENAH303684E6 Enamine

1-methyl-1H,4H,6H,7H-pyrano[4,3-c]pyrazole-3-carboxylic acid

  • MDL number MFCD28897873
  • Cas Number : 1533231-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1681046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1681046
Availability details for
Quantity
  • Availability
  • Add to Cart