ENAH303684E9 Enamine

6,8-dioxa-7lambda6-thiaspiro[3.5]nonane-7,7-dione

  • MDL number MFCD31719678
  • Cas Number : 2253631-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1682024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1682024
Availability details for
Quantity
  • Availability
  • Add to Cart