ENAH303684EC Enamine

2-(2,4-difluorophenyl)propanenitrile

  • MDL number MFCD11036492
  • Cas Number : 1096815-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1692372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1692372
Availability details for
Quantity
  • Availability
  • Add to Cart