ENAH303684FF Enamine

octahydropyrrolo[1,2-a]pyrazin-4-one

  • MDL number MFCD09878659
  • Cas Number : 1000577-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1696143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1696143
Availability details for
Quantity
  • Availability
  • Add to Cart