ENAH3036850A Enamine

5-difluoromethanesulfonyl-1-phenyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD31736468
  • Cas Number : 1219454-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-70
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1697923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1697923
Availability details for
Quantity
  • Availability
  • Add to Cart