ENAH3036850C Enamine

2-(2-fluorophenyl)propanenitrile

  • MDL number MFCD11036484
  • Cas Number : 1096308-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1698403

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1698403
Availability details for
Quantity
  • Availability
  • Add to Cart