ENAH30368511 Enamine

3-(4-bromophenyl)oxolane-3-carbaldehyde

  • MDL number MFCD31719508
  • Cas Number : 2253641-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1698689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1698689
Availability details for
Quantity
  • Availability
  • Add to Cart