ENAH30368514 Enamine

4-oxo-4H,5H,6H,7H,8H-thieno[3,2-c]azepine-2-carboxylic acid

  • MDL number MFCD31718888
  • Cas Number : 1501177-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1699013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1699013
Availability details for
Quantity
  • Availability
  • Add to Cart