ENAH30368515 Enamine

spiro[3.3]heptane-2-carbaldehyde

  • MDL number MFCD28977795
  • Cas Number : 1934935-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1478700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1478700
Availability details for
Quantity
  • Availability
  • Add to Cart