ENAH3036851B Enamine

2-(1,4-dioxan-2-yl)ethane-1-sulfonamide

  • MDL number MFCD31718889
  • Cas Number : 2248312-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1699658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1699658
Availability details for
Quantity
  • Availability
  • Add to Cart