ENAH31551611 Enamine

4-[2-(morpholin-4-yl)ethoxy]phenol

  • MDL number MFCD11181704
  • Cas Number : 141673-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58288
Availability details for
Quantity
  • Availability
  • Add to Cart