ENAH3155161B Enamine

5-(chloromethyl)-2-methoxypyridine hydrochloride

  • MDL number MFCD10697568
  • Cas Number : 120276-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85336
Availability details for
Quantity
  • Availability
  • Add to Cart