ENAH3155161C Enamine

N-{3-ethoxyspiro[3.5]nonan-1-ylidene}hydroxylamine

  • Cas Number : 1937361-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85527
Availability details for
Quantity
  • Availability
  • Add to Cart