ENAH31551622 Enamine

3-methyl-1,2-benzothiazole

  • MDL number MFCD18451373
  • Cas Number : 6187-89-9
  • Molecular Weight : 149.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94217

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-94217
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart